Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48401592-48401861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK8
Entrez Gene ID
5599
Gene Name
mitogen-activated protein kinase 8
Gene Aliases
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c
UniGene
Hs.138211
Gene Chromosome Location
Chr.10: 48306639-48439360 on Build GRCh38
Transcripts
NM_001278547.1 NM_001278548.1 NM_001323302.1 NM_001323320.1 NM_001323321.1 NM_001323322.1 NM_001323323.1 NM_001323324.1 NM_001323325.1 NM_001323326.1 NM_001323327.1 NM_001323328.1 NM_001323329.1 NM_001323330.1 NM_001323331.1 NM_139046.3 NM_139049.3 NR_136583.1 NR_136584.1 NR_136585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5352199 c.29_32delTTTA p.F10fs*3 Chr.10: 48401689-48401692 on Build GRCh38
5074514 c.16C>T p.R6C Chr.10: 48401676-48401676 on Build GRCh38
216920 c.74G>A p.R25Q Chr.10: 48401734-48401734 on Build GRCh38
918463 c.92C>T p.P31L Chr.10: 48401752-48401752 on Build GRCh38
7129307 c.92C>A p.P31H Chr.10: 48401752-48401752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773780784 Chr.10: 48401676-48401676 on Build GRCh38
rs777584404 Chr.10: 48401765-48401765 on Build GRCh38
rs746212436 Chr.10: 48401618-48401618 on Build GRCh38
rs769657154 Chr.10: 48401655-48401655 on Build GRCh38
rs778445166 Chr.10: 48401791-48401791 on Build GRCh38
rs774992035 Chr.10: 48401810-48401810 on Build GRCh38
rs745964104 Chr.10: 48401735-48401735 on Build GRCh38
rs751861105 Chr.10: 48401732-48401732 on Build GRCh38
rs755894470 Chr.10: 48401734-48401734 on Build GRCh38
rs370411166 Chr.10: 48401724-48401724 on Build GRCh38
rs947509151 Chr.10: 48401652-48401652 on Build GRCh38
rs745375804 Chr.10: 48401793-48401793 on Build GRCh38
rs749289588 Chr.10: 48401660-48401660 on Build GRCh38
rs771699494 Chr.10: 48401797-48401797 on Build GRCh38
rs137900446 Chr.10: 48401708-48401708 on Build GRCh38
rs750494171 Chr.10: 48401723-48401723 on Build GRCh38
rs777670645 Chr.10: 48401656-48401656 on Build GRCh38
rs886105474 Chr.10: 48401777-48401777 on Build GRCh38
rs200375054 Chr.10: 48401618-48401618 on Build GRCh38
rs150753150 Chr.10: 48401687-48401687 on Build GRCh38
rs764612003 Chr.10: 48401700-48401700 on Build GRCh38
rs779204208 Chr.10: 48401619-48401619 on Build GRCh38
rs759178320 Chr.10: 48401677-48401677 on Build GRCh38
rs747898436 Chr.10: 48401654-48401654 on Build GRCh38
rs754396010 Chr.10: 48401701-48401701 on Build GRCh38
rs770874861 Chr.10: 48401782-48401782 on Build GRCh38
rs760472627 Chr.10: 48401811-48401811 on Build GRCh38
rs1047240487 Chr.10: 48401686-48401686 on Build GRCh38
rs771655862 Chr.10: 48401683-48401683 on Build GRCh38
rs1016170027 Chr.10: 48401828-48401828 on Build GRCh38
rs199529538 Chr.10: 48401639-48401639 on Build GRCh38
rs760960935 Chr.10: 48401691-48401691 on Build GRCh38
rs780150000 Chr.10: 48401731-48401731 on Build GRCh38
rs149467709 Chr.10: 48401768-48401768 on Build GRCh38
rs780993770 Chr.10: 48401651-48401651 on Build GRCh38
rs201739023 Chr.10: 48401826-48401826 on Build GRCh38
rs920199360 Chr.10: 48401627-48401627 on Build GRCh38
rs765388602 Chr.10: 48401715-48401715 on Build GRCh38
rs770396528 Chr.10: 48401665-48401665 on Build GRCh38

View Full Product Details