Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179201167-179201436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3CA
Entrez Gene ID
5290
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha
Gene Aliases
CLOVE, CWS5, MCAP, MCM, MCMTC, PI3K, PI3K-alpha, p110-alpha
UniGene
Hs.553498
Gene Chromosome Location
Chr.3: 179148114-179240093 on Build GRCh38
Transcripts
NM_006218.3 XM_006713658.3 XM_011512894.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3695924 c.670G>T p.A224S Chr.3: 179201397-179201397 on Build GRCh38
6189010 c.584G>A p.W195* Chr.3: 179201311-179201311 on Build GRCh38
6475749 c.638A>C p.H213P Chr.3: 179201365-179201365 on Build GRCh38
6964034 c.596C>G p.S199C Chr.3: 179201323-179201323 on Build GRCh38
5398692 c.678G>C p.R226S Chr.3: 179201405-179201405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749008800 Chr.3: 179201248-179201251 on Build GRCh38
rs751662352 Chr.3: 179201362-179201362 on Build GRCh38
rs535774183 Chr.3: 179201236-179201236 on Build GRCh38
rs775151613 Chr.3: 179201256-179201256 on Build GRCh38
rs769993336 Chr.3: 179201231-179201231 on Build GRCh38
rs758698734 Chr.3: 179201251-179201254 on Build GRCh38
rs202158911 Chr.3: 179201303-179201303 on Build GRCh38
rs587777792 Chr.3: 179201379-179201379 on Build GRCh38
rs747466381 Chr.3: 179201261-179201261 on Build GRCh38
rs541134560 Chr.3: 179201313-179201313 on Build GRCh38
rs919212078 Chr.3: 179201268-179201268 on Build GRCh38
rs200507422 Chr.3: 179201287-179201287 on Build GRCh38
rs776828704 Chr.3: 179201276-179201276 on Build GRCh38
rs910287504 Chr.3: 179201373-179201373 on Build GRCh38
rs375704434 Chr.3: 179201218-179201218 on Build GRCh38
rs200131834 Chr.3: 179201341-179201341 on Build GRCh38
rs769316233 Chr.3: 179201267-179201267 on Build GRCh38
rs943131817 Chr.3: 179201395-179201395 on Build GRCh38
rs763090975 Chr.3: 179201325-179201325 on Build GRCh38
rs933769772 Chr.3: 179201247-179201247 on Build GRCh38
rs773550451 Chr.3: 179201318-179201318 on Build GRCh38
rs923421083 Chr.3: 179201203-179201203 on Build GRCh38
rs372610277 Chr.3: 179201350-179201350 on Build GRCh38

View Full Product Details