Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97733432-97733659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
UniGene
Hs.234569
Gene Chromosome Location
Chr.2: 97712030-97744327 on Build GRCh38
Transcripts
NM_001079.3 XM_017004867.1 XM_017004868.1 XM_017004869.1 XM_017004870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5736138 c.847C>T p.R283* Chr.2: 97733553-97733553 on Build GRCh38
6824752 c.474C>T p.I158I Chr.2: 97733558-97733558 on Build GRCh38
4412601 c.889+6C>T p.? Chr.2: 97733601-97733601 on Build GRCh38
1233086 c.853G>A p.D285N Chr.2: 97733559-97733559 on Build GRCh38
5736139 c.469C>T p.R157* Chr.2: 97733553-97733553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112357205 Chr.2: 97733493-97733493 on Build GRCh38
rs553261328 Chr.2: 97733558-97733558 on Build GRCh38
rs909677452 Chr.2: 97733584-97733584 on Build GRCh38
rs982143659 Chr.2: 97733480-97733480 on Build GRCh38
rs202079520 Chr.2: 97733505-97733505 on Build GRCh38
rs774658245 Chr.2: 97733603-97733603 on Build GRCh38
rs200585997 Chr.2: 97733512-97733512 on Build GRCh38
rs746378071 Chr.2: 97733579-97733579 on Build GRCh38
rs536451792 Chr.2: 97733527-97733527 on Build GRCh38
rs747446900 Chr.2: 97733459-97733459 on Build GRCh38
rs761700835 Chr.2: 97733520-97733520 on Build GRCh38
rs993494098 Chr.2: 97733633-97733633 on Build GRCh38
rs773298062 Chr.2: 97733539-97733539 on Build GRCh38
rs201457490 Chr.2: 97733601-97733601 on Build GRCh38
rs769875156 Chr.2: 97733486-97733486 on Build GRCh38
rs56096305 Chr.2: 97733602-97733602 on Build GRCh38
rs201810987 Chr.2: 97733617-97733617 on Build GRCh38
rs746322954 Chr.2: 97733482-97733482 on Build GRCh38
rs781685153 Chr.2: 97733475-97733475 on Build GRCh38
rs771165725 Chr.2: 97733474-97733474 on Build GRCh38
rs544647865 Chr.2: 97733615-97733615 on Build GRCh38
rs756672820 Chr.2: 97733585-97733585 on Build GRCh38
rs200501678 Chr.2: 97733641-97733641 on Build GRCh38
rs754420169 Chr.2: 97733553-97733553 on Build GRCh38
rs764977425 Chr.2: 97733554-97733554 on Build GRCh38
rs773211731 Chr.2: 97733610-97733610 on Build GRCh38
rs370478398 Chr.2: 97733498-97733498 on Build GRCh38
rs113994173 Chr.2: 97733464-97733464 on Build GRCh38
rs781477800 Chr.2: 97733559-97733559 on Build GRCh38
rs774406382 Chr.2: 97733508-97733508 on Build GRCh38
rs760529841 Chr.2: 97733614-97733614 on Build GRCh38
rs200437787 Chr.2: 97733626-97733626 on Build GRCh38
rs370811702 Chr.2: 97733607-97733607 on Build GRCh38
rs201824457 Chr.2: 97733620-97733620 on Build GRCh38
rs753453013 Chr.2: 97733548-97733548 on Build GRCh38
rs780517718 Chr.2: 97733589-97733589 on Build GRCh38
rs200126170 Chr.2: 97733572-97733572 on Build GRCh38
rs56081234 Chr.2: 97733638-97733638 on Build GRCh38
rs201225206 Chr.2: 97733592-97733592 on Build GRCh38
rs897858343 Chr.2: 97733618-97733618 on Build GRCh38
rs760626189 Chr.2: 97733544-97733544 on Build GRCh38
rs557179479 Chr.2: 97733506-97733506 on Build GRCh38
rs759115124 Chr.2: 97733619-97733619 on Build GRCh38
rs201017686 Chr.2: 97733609-97733609 on Build GRCh38
rs200987919 Chr.2: 97733632-97733632 on Build GRCh38

View Full Product Details