Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42713163-42713394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC13
Entrez Gene ID
152206
Gene Name
coiled-coil domain containing 13
Gene Aliases
-
UniGene
Hs.604182
Gene Chromosome Location
Chr.3: 42704118-42773253 on Build GRCh38
Transcripts
NM_144719.3 XM_005264898.3 XM_011533418.2 XM_011533419.2 XM_017005781.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1485184 c.1788G>A p.V596V Chr.3: 42713247-42713247 on Build GRCh38
3974413 c.1775G>A p.R592Q Chr.3: 42713260-42713260 on Build GRCh38
4917824 c.1782C>T p.R594R Chr.3: 42713253-42713253 on Build GRCh38
446517 c.1755G>C p.E585D Chr.3: 42713280-42713280 on Build GRCh38
5471730 c.1774C>T p.R592* Chr.3: 42713261-42713261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036743889 Chr.3: 42713260-42713260 on Build GRCh38
rs765582369 Chr.3: 42713289-42713289 on Build GRCh38
rs984663349 Chr.3: 42713255-42713255 on Build GRCh38
rs916819488 Chr.3: 42713182-42713182 on Build GRCh38
rs777554897 Chr.3: 42713326-42713326 on Build GRCh38
rs765845945 Chr.3: 42713249-42713250 on Build GRCh38
rs187019100 Chr.3: 42713335-42713335 on Build GRCh38
rs777628129 Chr.3: 42713248-42713248 on Build GRCh38
rs749211540 Chr.3: 42713332-42713332 on Build GRCh38
rs765734392 Chr.3: 42713210-42713210 on Build GRCh38
rs754506293 Chr.3: 42713309-42713309 on Build GRCh38
rs750922481 Chr.3: 42713250-42713251 on Build GRCh38
rs775504311 Chr.3: 42713254-42713254 on Build GRCh38
rs991074004 Chr.3: 42713370-42713370 on Build GRCh38
rs1049662284 Chr.3: 42713268-42713268 on Build GRCh38
rs751289042 Chr.3: 42713221-42713221 on Build GRCh38
rs754253597 Chr.3: 42713184-42713184 on Build GRCh38
rs758030012 Chr.3: 42713278-42713281 on Build GRCh38
rs762144812 Chr.3: 42713271-42713271 on Build GRCh38
rs889682321 Chr.3: 42713298-42713298 on Build GRCh38
rs777796591 Chr.3: 42713226-42713226 on Build GRCh38
rs930987050 Chr.3: 42713267-42713267 on Build GRCh38
rs767345602 Chr.3: 42713310-42713310 on Build GRCh38
rs143078504 Chr.3: 42713201-42713201 on Build GRCh38
rs763478190 Chr.3: 42713305-42713305 on Build GRCh38
rs376881127 Chr.3: 42713363-42713363 on Build GRCh38
rs373122697 Chr.3: 42713336-42713336 on Build GRCh38
rs928338594 Chr.3: 42713347-42713347 on Build GRCh38
rs747162398 Chr.3: 42713353-42713353 on Build GRCh38
rs757861196 Chr.3: 42713218-42713218 on Build GRCh38
rs970906496 Chr.3: 42713193-42713193 on Build GRCh38
rs755874084 Chr.3: 42713319-42713319 on Build GRCh38
rs901736657 Chr.3: 42713189-42713189 on Build GRCh38
rs1040870854 Chr.3: 42713321-42713321 on Build GRCh38
rs1008077145 Chr.3: 42713304-42713304 on Build GRCh38
rs776838896 Chr.3: 42713264-42713264 on Build GRCh38
rs747995879 Chr.3: 42713234-42713234 on Build GRCh38
rs757165954 Chr.3: 42713334-42713334 on Build GRCh38
rs996091377 Chr.3: 42713208-42713208 on Build GRCh38
rs754629090 Chr.3: 42713222-42713222 on Build GRCh38
rs755931457 Chr.3: 42713244-42713244 on Build GRCh38
rs369481975 Chr.3: 42713242-42713242 on Build GRCh38
rs750931943 Chr.3: 42713214-42713214 on Build GRCh38
rs145418945 Chr.3: 42713219-42713219 on Build GRCh38
rs752382670 Chr.3: 42713312-42713312 on Build GRCh38
rs754217368 Chr.3: 42713205-42713205 on Build GRCh38
rs537764027 Chr.3: 42713308-42713308 on Build GRCh38
rs768909971 Chr.3: 42713261-42713261 on Build GRCh38
rs778795529 Chr.3: 42713341-42713341 on Build GRCh38
rs749188442 Chr.3: 42713249-42713249 on Build GRCh38
rs773598757 Chr.3: 42713300-42713300 on Build GRCh38

View Full Product Details