Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41332117-41332378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFB1
Entrez Gene ID
7040
Gene Name
transforming growth factor beta 1
Gene Aliases
CED, DPD1, LAP, TGFB, TGFbeta
UniGene
Hs.645227
Gene Chromosome Location
Chr.19: 41330531-41353933 on Build GRCh38
Transcripts
NM_000660.5 XM_011527242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078502 c.914A>G p.D305G Chr.19: 41332228-41332228 on Build GRCh38
6869186 c.969C>T p.L323L Chr.19: 41332173-41332173 on Build GRCh38
6049577 c.873G>A p.K291K Chr.19: 41332269-41332269 on Build GRCh38
6800692 c.883G>A p.V295M Chr.19: 41332259-41332259 on Build GRCh38
6800691 c.937G>A p.E313K Chr.19: 41332205-41332205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200763912 Chr.19: 41332203-41332203 on Build GRCh38
rs745482429 Chr.19: 41332151-41332151 on Build GRCh38
rs776406237 Chr.19: 41332173-41332173 on Build GRCh38
rs775861573 Chr.19: 41332234-41332234 on Build GRCh38
rs761032649 Chr.19: 41332306-41332306 on Build GRCh38
rs190566789 Chr.19: 41332269-41332269 on Build GRCh38
rs201281623 Chr.19: 41332149-41332149 on Build GRCh38
rs759582721 Chr.19: 41332200-41332200 on Build GRCh38
rs761279576 Chr.19: 41332220-41332220 on Build GRCh38
rs199516461 Chr.19: 41332198-41332198 on Build GRCh38
rs770754343 Chr.19: 41332276-41332276 on Build GRCh38
rs200527282 Chr.19: 41332235-41332235 on Build GRCh38
rs200164212 Chr.19: 41332255-41332255 on Build GRCh38
rs199946261 Chr.19: 41332226-41332226 on Build GRCh38
rs779818818 Chr.19: 41332248-41332248 on Build GRCh38
rs72480429 Chr.19: 41332224-41332224 on Build GRCh38
rs748135261 Chr.19: 41332175-41332175 on Build GRCh38
rs11466358 Chr.19: 41332170-41332170 on Build GRCh38
rs200209614 Chr.19: 41332169-41332169 on Build GRCh38
rs747968669 Chr.19: 41332270-41332270 on Build GRCh38
rs201366321 Chr.19: 41332185-41332185 on Build GRCh38
rs768250306 Chr.19: 41332172-41332172 on Build GRCh38
rs767685429 Chr.19: 41332206-41332206 on Build GRCh38
rs199713772 Chr.19: 41332150-41332150 on Build GRCh38
rs573968848 Chr.19: 41332352-41332352 on Build GRCh38
rs753287325 Chr.19: 41332141-41332141 on Build GRCh38
rs11551225 Chr.19: 41332179-41332179 on Build GRCh38
rs758130535 Chr.19: 41332164-41332164 on Build GRCh38
rs774427089 Chr.19: 41332267-41332269 on Build GRCh38
rs201635147 Chr.19: 41332243-41332243 on Build GRCh38
rs759827699 Chr.19: 41332300-41332300 on Build GRCh38
rs199952420 Chr.19: 41332314-41332314 on Build GRCh38
rs769782160 Chr.19: 41332275-41332275 on Build GRCh38
rs56361919 Chr.19: 41332163-41332163 on Build GRCh38
rs763101036 Chr.19: 41332189-41332189 on Build GRCh38
rs544266201 Chr.19: 41332358-41332358 on Build GRCh38
rs769434404 Chr.19: 41332181-41332181 on Build GRCh38
rs139977837 Chr.19: 41332250-41332250 on Build GRCh38
rs8179181 Chr.19: 41332301-41332301 on Build GRCh38
rs369182751 Chr.19: 41332216-41332216 on Build GRCh38
rs200230083 Chr.19: 41332145-41332145 on Build GRCh38
rs200839435 Chr.19: 41332298-41332298 on Build GRCh38
rs201019135 Chr.19: 41332322-41332322 on Build GRCh38
rs1027898259 Chr.19: 41332303-41332303 on Build GRCh38
rs766572720 Chr.19: 41332197-41332197 on Build GRCh38

View Full Product Details