Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63579982-63580253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTP4A1
Entrez Gene ID
7803
Gene Name
protein tyrosine phosphatase type IVA, member 1
Gene Aliases
HH72, PRL-1, PRL1, PTP(CAAX1), PTPCAAX1
UniGene
Hs.227777
Gene Chromosome Location
Chr.6: 63528021-63583588 on Build GRCh38
Transcripts
NM_003463.4 XM_011536111.1 XM_011536112.1 XM_017011270.1 XM_017011271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5634022 c.439C>T p.L147L Chr.6: 63580091-63580091 on Build GRCh38
6336013 c.441G>T p.L147L Chr.6: 63580093-63580093 on Build GRCh38
3160221 c.458G>A p.R153H Chr.6: 63580110-63580110 on Build GRCh38
5768138 c.450G>A p.E150E Chr.6: 63580102-63580102 on Build GRCh38
6201011 c.405-8C>T p.? Chr.6: 63580049-63580049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62412934 Chr.6: 63580050-63580050 on Build GRCh38
rs199682183 Chr.6: 63580083-63580083 on Build GRCh38
rs546134768 Chr.6: 63580145-63580145 on Build GRCh38
rs754493391 Chr.6: 63580065-63580065 on Build GRCh38
rs747957784 Chr.6: 63580033-63580034 on Build GRCh38
rs947354115 Chr.6: 63580188-63580188 on Build GRCh38
rs368017955 Chr.6: 63580123-63580123 on Build GRCh38
rs77267139 Chr.6: 63580048-63580048 on Build GRCh38
rs753655665 Chr.6: 63580207-63580207 on Build GRCh38
rs528551398 Chr.6: 63580206-63580206 on Build GRCh38
rs951194134 Chr.6: 63580028-63580028 on Build GRCh38
rs773911774 Chr.6: 63580176-63580176 on Build GRCh38
rs56188808 Chr.6: 63580048-63580048 on Build GRCh38
rs776990512 Chr.6: 63580219-63580219 on Build GRCh38
rs575722475 Chr.6: 63580130-63580130 on Build GRCh38
rs141880918 Chr.6: 63580144-63580144 on Build GRCh38
rs777163209 Chr.6: 63580162-63580162 on Build GRCh38
rs866386335 Chr.6: 63580078-63580078 on Build GRCh38
rs764372362 Chr.6: 63580195-63580195 on Build GRCh38
rs2758266 Chr.6: 63580026-63580026 on Build GRCh38
rs563964139 Chr.6: 63580110-63580110 on Build GRCh38
rs754947851 Chr.6: 63580034-63580035 on Build GRCh38
rs774841816 Chr.6: 63580034-63580034 on Build GRCh38
rs1618240 Chr.6: 63580063-63580063 on Build GRCh38
rs766997007 Chr.6: 63580179-63580179 on Build GRCh38
rs779059610 Chr.6: 63580128-63580128 on Build GRCh38
rs201632872 Chr.6: 63580171-63580171 on Build GRCh38
rs201134694 Chr.6: 63580215-63580215 on Build GRCh38
rs752197709 Chr.6: 63580184-63580184 on Build GRCh38
rs200202391 Chr.6: 63580205-63580205 on Build GRCh38
rs778739544 Chr.6: 63580220-63580220 on Build GRCh38
rs769218125 Chr.6: 63580157-63580157 on Build GRCh38
rs548081382 Chr.6: 63580105-63580105 on Build GRCh38
rs757095424 Chr.6: 63580210-63580210 on Build GRCh38
rs757501922 Chr.6: 63580127-63580127 on Build GRCh38
rs756070386 Chr.6: 63580108-63580108 on Build GRCh38
rs759225817 Chr.6: 63580178-63580178 on Build GRCh38
rs772109649 Chr.6: 63580143-63580143 on Build GRCh38
rs1681943 Chr.6: 63580091-63580091 on Build GRCh38
rs758129954 Chr.6: 63580034-63580036 on Build GRCh38
rs762214973 Chr.6: 63580166-63580166 on Build GRCh38
rs921517969 Chr.6: 63580164-63580164 on Build GRCh38
rs747484192 Chr.6: 63580156-63580156 on Build GRCh38

View Full Product Details