Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47637914-47638151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK1
Entrez Gene ID
2002
Gene Name
ELK1, ETS transcription factor
Gene Aliases
-
UniGene
Hs.181128
Gene Chromosome Location
Chr.X: 47635520-47650604 on Build GRCh38
Transcripts
NM_001114123.2 NM_001257168.1 NM_005229.4 XM_017029339.1

SNP Detail

SNP ID Chromosome Location
rs755114172 Chr.X: 47638007-47638007 on Build GRCh38
rs369027699 Chr.X: 47637967-47637967 on Build GRCh38
rs376822462 Chr.X: 47638112-47638112 on Build GRCh38
rs200172068 Chr.X: 47638009-47638009 on Build GRCh38
rs778250617 Chr.X: 47638110-47638110 on Build GRCh38
rs773489927 Chr.X: 47637990-47637990 on Build GRCh38
rs751772833 Chr.X: 47638001-47638001 on Build GRCh38
rs752423073 Chr.X: 47638027-47638027 on Build GRCh38
rs756731775 Chr.X: 47638102-47638102 on Build GRCh38
rs371259584 Chr.X: 47638113-47638113 on Build GRCh38
rs200141730 Chr.X: 47638124-47638124 on Build GRCh38
rs766556938 Chr.X: 47637995-47637995 on Build GRCh38
rs376525319 Chr.X: 47637993-47637993 on Build GRCh38
rs777459618 Chr.X: 47637941-47637941 on Build GRCh38
rs747375894 Chr.X: 47637962-47637962 on Build GRCh38
rs765768273 Chr.X: 47637975-47637975 on Build GRCh38
rs762419349 Chr.X: 47637973-47637973 on Build GRCh38
rs774035251 Chr.X: 47637947-47637947 on Build GRCh38
rs201698802 Chr.X: 47638057-47638057 on Build GRCh38
rs766899855 Chr.X: 47638123-47638123 on Build GRCh38
rs769100410 Chr.X: 47637965-47637965 on Build GRCh38
rs756113016 Chr.X: 47638053-47638053 on Build GRCh38
rs373267605 Chr.X: 47637974-47637974 on Build GRCh38

View Full Product Details