Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41341857-41342126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFB1
Entrez Gene ID
7040
Gene Name
transforming growth factor beta 1
Gene Aliases
CED, DPD1, LAP, TGFB, TGFbeta
UniGene
Hs.645227
Gene Chromosome Location
Chr.19: 41330531-41353933 on Build GRCh38
Transcripts
NM_000660.5 XM_011527242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6084917 c.860+1G>T p.? Chr.19: 41341882-41341882 on Build GRCh38
1684582 c.713-9delC p.? Chr.19: 41342039-41342039 on Build GRCh38
6457611 c.806A>G p.Q269R Chr.19: 41341937-41341937 on Build GRCh38
1288443 c.785C>A p.A262D Chr.19: 41341958-41341958 on Build GRCh38
1229076 c.784G>A p.A262T Chr.19: 41341959-41341959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547881966 Chr.19: 41341962-41341962 on Build GRCh38
rs1025241895 Chr.19: 41341906-41341906 on Build GRCh38
rs149900977 Chr.19: 41341979-41341979 on Build GRCh38
rs778626011 Chr.19: 41341995-41341995 on Build GRCh38
rs746014542 Chr.19: 41341909-41341909 on Build GRCh38
rs199803251 Chr.19: 41342056-41342056 on Build GRCh38
rs201119217 Chr.19: 41341912-41341912 on Build GRCh38
rs757193903 Chr.19: 41341994-41341994 on Build GRCh38
rs749275172 Chr.19: 41341892-41341892 on Build GRCh38
rs72480428 Chr.19: 41342037-41342037 on Build GRCh38
rs747216224 Chr.19: 41342022-41342022 on Build GRCh38
rs763345073 Chr.19: 41342051-41342051 on Build GRCh38
rs201948484 Chr.19: 41341935-41341935 on Build GRCh38
rs373586759 Chr.19: 41341942-41341942 on Build GRCh38
rs569594975 Chr.19: 41342028-41342028 on Build GRCh38
rs760991789 Chr.19: 41342065-41342066 on Build GRCh38
rs766703879 Chr.19: 41342054-41342054 on Build GRCh38
rs772594713 Chr.19: 41342050-41342051 on Build GRCh38
rs72480424 Chr.19: 41342021-41342021 on Build GRCh38
rs755630382 Chr.19: 41341963-41341963 on Build GRCh38
rs377448414 Chr.19: 41342049-41342049 on Build GRCh38
rs1038400154 Chr.19: 41341933-41341933 on Build GRCh38
rs767192967 Chr.19: 41341958-41341958 on Build GRCh38
rs35383147 Chr.19: 41342020-41342020 on Build GRCh38
rs973558315 Chr.19: 41341978-41341978 on Build GRCh38
rs141484614 Chr.19: 41341924-41341924 on Build GRCh38
rs764932015 Chr.19: 41342075-41342076 on Build GRCh38
rs778711968 Chr.19: 41341904-41341904 on Build GRCh38
rs546927712 Chr.19: 41342014-41342014 on Build GRCh38
rs530534755 Chr.19: 41342033-41342033 on Build GRCh38
rs267605501 Chr.19: 41341945-41341945 on Build GRCh38
rs775636009 Chr.19: 41341922-41341922 on Build GRCh38
rs55659002 Chr.19: 41342038-41342038 on Build GRCh38
rs200558128 Chr.19: 41341951-41341951 on Build GRCh38
rs985692714 Chr.19: 41341894-41341894 on Build GRCh38
rs368253972 Chr.19: 41342076-41342076 on Build GRCh38
rs559265663 Chr.19: 41341925-41341925 on Build GRCh38
rs1056983716 Chr.19: 41341954-41341954 on Build GRCh38
rs148552731 Chr.19: 41342025-41342025 on Build GRCh38
rs761896338 Chr.19: 41342099-41342099 on Build GRCh38
rs34233206 Chr.19: 41342011-41342012 on Build GRCh38
rs112108984 Chr.19: 41342032-41342032 on Build GRCh38
rs371563202 Chr.19: 41341878-41341878 on Build GRCh38
rs146182311 Chr.19: 41341952-41341952 on Build GRCh38
rs745664232 Chr.19: 41342019-41342019 on Build GRCh38
rs773593055 Chr.19: 41342066-41342069 on Build GRCh38
rs767516971 Chr.19: 41342100-41342100 on Build GRCh38
rs1800472 Chr.19: 41341955-41341955 on Build GRCh38
rs570538520 Chr.19: 41342106-41342106 on Build GRCh38
rs139080695 Chr.19: 41342065-41342065 on Build GRCh38
rs770664280 Chr.19: 41341897-41341897 on Build GRCh38
rs753735387 Chr.19: 41341980-41341980 on Build GRCh38
rs28365167 Chr.19: 41342038-41342038 on Build GRCh38
rs943251438 Chr.19: 41342053-41342053 on Build GRCh38
rs918952473 Chr.19: 41341991-41341991 on Build GRCh38

View Full Product Details