Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42735634-42735901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC13
Entrez Gene ID
152206
Gene Name
coiled-coil domain containing 13
Gene Aliases
-
UniGene
Hs.604182
Gene Chromosome Location
Chr.3: 42704118-42773253 on Build GRCh38
Transcripts
NM_144719.3 XM_005264898.3 XM_005264899.4 XM_011533418.2 XM_011533419.2 XM_011533420.2 XM_011533421.2 XM_017005781.1 NR_046732.1

Gene Symbol
CCDC13-AS1
Entrez Gene ID
100874114
Gene Name
CCDC13 antisense RNA 1
Gene Aliases
-
UniGene
Hs.669654
Gene Chromosome Location
Chr.3: 42732575-42745158 on Build GRCh38
Transcripts
NM_144719.3 XM_005264898.3 XM_005264899.4 XM_011533418.2 XM_011533419.2 XM_011533420.2 XM_011533421.2 XM_017005781.1 NR_046732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199820 c.1274G>A p.R425Q Chr.3: 42735804-42735804 on Build GRCh38
3916057 c.1190T>G p.I397S Chr.3: 42735888-42735888 on Build GRCh38
5556592 c.1371C>T p.H457H Chr.3: 42735707-42735707 on Build GRCh38
6441482 c.1355G>A p.G452E Chr.3: 42735723-42735723 on Build GRCh38
1485185 c.1317G>A p.E439E Chr.3: 42735761-42735761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368238525 Chr.3: 42735656-42735656 on Build GRCh38
rs993642023 Chr.3: 42735823-42735823 on Build GRCh38
rs777005407 Chr.3: 42735805-42735805 on Build GRCh38
rs565579778 Chr.3: 42735686-42735686 on Build GRCh38
rs749871481 Chr.3: 42735772-42735772 on Build GRCh38
rs775680333 Chr.3: 42735795-42735795 on Build GRCh38
rs745463382 Chr.3: 42735679-42735679 on Build GRCh38
rs376795920 Chr.3: 42735760-42735760 on Build GRCh38
rs768754276 Chr.3: 42735691-42735691 on Build GRCh38
rs756126098 Chr.3: 42735851-42735851 on Build GRCh38
rs766193108 Chr.3: 42735818-42735818 on Build GRCh38
rs746867668 Chr.3: 42735759-42735759 on Build GRCh38
rs761082722 Chr.3: 42735796-42735796 on Build GRCh38
rs754354098 Chr.3: 42735716-42735716 on Build GRCh38
rs867459450 Chr.3: 42735657-42735657 on Build GRCh38
rs369300600 Chr.3: 42735787-42735787 on Build GRCh38
rs564647857 Chr.3: 42735745-42735745 on Build GRCh38
rs751201115 Chr.3: 42735819-42735819 on Build GRCh38
rs780647548 Chr.3: 42735860-42735862 on Build GRCh38
rs778229694 Chr.3: 42735659-42735659 on Build GRCh38
rs201862149 Chr.3: 42735724-42735724 on Build GRCh38
rs746011265 Chr.3: 42735854-42735854 on Build GRCh38
rs748056672 Chr.3: 42735849-42735849 on Build GRCh38
rs772319974 Chr.3: 42735794-42735794 on Build GRCh38
rs528985670 Chr.3: 42735811-42735811 on Build GRCh38
rs896669774 Chr.3: 42735817-42735817 on Build GRCh38
rs780999456 Chr.3: 42735845-42735845 on Build GRCh38
rs745924733 Chr.3: 42735790-42735790 on Build GRCh38
rs778485573 Chr.3: 42735676-42735676 on Build GRCh38
rs114512148 Chr.3: 42735748-42735748 on Build GRCh38
rs760992984 Chr.3: 42735710-42735710 on Build GRCh38
rs377015472 Chr.3: 42735666-42735666 on Build GRCh38
rs770986343 Chr.3: 42735695-42735695 on Build GRCh38
rs764380663 Chr.3: 42735804-42735804 on Build GRCh38
rs200926180 Chr.3: 42735779-42735779 on Build GRCh38
rs1002443839 Chr.3: 42735806-42735806 on Build GRCh38
rs747977701 Chr.3: 42735875-42735876 on Build GRCh38
rs967884855 Chr.3: 42735714-42735714 on Build GRCh38
rs774439094 Chr.3: 42735702-42735702 on Build GRCh38
rs58970917 Chr.3: 42735877-42735877 on Build GRCh38
rs148765643 Chr.3: 42735855-42735855 on Build GRCh38
rs538648897 Chr.3: 42735814-42735814 on Build GRCh38
rs551175692 Chr.3: 42735707-42735707 on Build GRCh38
rs746783392 Chr.3: 42735687-42735687 on Build GRCh38
rs754827277 Chr.3: 42735839-42735839 on Build GRCh38
rs956450866 Chr.3: 42735744-42735744 on Build GRCh38
rs199592393 Chr.3: 42735665-42735665 on Build GRCh38
rs757981259 Chr.3: 42735815-42735815 on Build GRCh38
rs776704914 Chr.3: 42735693-42735693 on Build GRCh38
rs764648124 Chr.3: 42735712-42735712 on Build GRCh38
rs750067488 Chr.3: 42735725-42735725 on Build GRCh38
rs749388057 Chr.3: 42735694-42735694 on Build GRCh38
rs531063281 Chr.3: 42735706-42735706 on Build GRCh38
rs762335677 Chr.3: 42735810-42735810 on Build GRCh38
rs566513960 Chr.3: 42735788-42735788 on Build GRCh38
rs201124720 Chr.3: 42735798-42735798 on Build GRCh38
rs756728996 Chr.3: 42735717-42735717 on Build GRCh38
rs558541969 Chr.3: 42735852-42735852 on Build GRCh38

View Full Product Details