Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31575721-31575962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNF
Entrez Gene ID
7124
Gene Name
tumor necrosis factor
Gene Aliases
DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
UniGene
Hs.241570
Gene Chromosome Location
Chr.6: 31575567-31578336 on Build GRCh38
Transcripts
NM_000594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624579 c.21C>T p.I7I Chr.6: 31575762-31575762 on Build GRCh38
4734841 c.130G>A p.A44T Chr.6: 31575871-31575871 on Build GRCh38
3374203 c.99C>T p.L33L Chr.6: 31575840-31575840 on Build GRCh38
5560328 c.3G>A p.M1I Chr.6: 31575744-31575744 on Build GRCh38
4610793 c.40G>A p.E14K Chr.6: 31575781-31575781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3179060 Chr.6: 31575895-31575895 on Build GRCh38
rs144480538 Chr.6: 31575879-31575879 on Build GRCh38
rs576621666 Chr.6: 31575827-31575827 on Build GRCh38
rs377627218 Chr.6: 31575768-31575768 on Build GRCh38
rs779870893 Chr.6: 31575825-31575825 on Build GRCh38
rs764955335 Chr.6: 31575814-31575814 on Build GRCh38
rs753713562 Chr.6: 31575805-31575805 on Build GRCh38
rs771650207 Chr.6: 31575755-31575755 on Build GRCh38
rs777874746 Chr.6: 31575910-31575910 on Build GRCh38
rs541347219 Chr.6: 31575882-31575882 on Build GRCh38
rs765178016 Chr.6: 31575926-31575926 on Build GRCh38
rs747694851 Chr.6: 31575754-31575754 on Build GRCh38
rs565862537 Chr.6: 31575756-31575756 on Build GRCh38
rs867435711 Chr.6: 31575871-31575871 on Build GRCh38
rs953747858 Chr.6: 31575751-31575751 on Build GRCh38
rs369163834 Chr.6: 31575870-31575870 on Build GRCh38
rs948780599 Chr.6: 31575875-31575875 on Build GRCh38
rs754639425 Chr.6: 31575811-31575811 on Build GRCh38
rs201502336 Chr.6: 31575764-31575764 on Build GRCh38
rs377338702 Chr.6: 31575797-31575797 on Build GRCh38
rs982610810 Chr.6: 31575766-31575766 on Build GRCh38
rs369780852 Chr.6: 31575743-31575743 on Build GRCh38
rs759378982 Chr.6: 31575924-31575924 on Build GRCh38
rs141667614 Chr.6: 31575861-31575861 on Build GRCh38
rs368044930 Chr.6: 31575934-31575934 on Build GRCh38
rs757794184 Chr.6: 31575853-31575853 on Build GRCh38
rs775506502 Chr.6: 31575781-31575781 on Build GRCh38
rs558678940 Chr.6: 31575862-31575862 on Build GRCh38
rs756182468 Chr.6: 31575902-31575902 on Build GRCh38
rs766034823 Chr.6: 31575804-31575804 on Build GRCh38
rs2228088 Chr.6: 31575828-31575828 on Build GRCh38
rs1036935762 Chr.6: 31575880-31575880 on Build GRCh38
rs770023902 Chr.6: 31575771-31575771 on Build GRCh38
rs759696973 Chr.6: 31575769-31575769 on Build GRCh38
rs1049309348 Chr.6: 31575867-31575868 on Build GRCh38
rs972459518 Chr.6: 31575784-31575784 on Build GRCh38
rs374531985 Chr.6: 31575763-31575763 on Build GRCh38
rs777405140 Chr.6: 31575836-31575836 on Build GRCh38
rs146375573 Chr.6: 31575789-31575789 on Build GRCh38
rs915225183 Chr.6: 31575842-31575842 on Build GRCh38
rs776753689 Chr.6: 31575912-31575912 on Build GRCh38
rs200241887 Chr.6: 31575878-31575878 on Build GRCh38
rs747406665 Chr.6: 31575838-31575838 on Build GRCh38

View Full Product Details