Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 175097223-175097490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF2
Entrez Gene ID
1386
Gene Name
activating transcription factor 2
Gene Aliases
CRE-BP1, CREB-2, CREB2, HB16, TREB7
UniGene
Hs.592510
Gene Chromosome Location
Chr.2: 175072250-175168206 on Build GRCh38
Transcripts
NM_001256090.1 NM_001256091.1 NM_001256092.1 NM_001256093.1 NM_001880.3 NR_045768.1 NR_045769.1 NR_045770.1 NR_045771.1 NR_045772.1 NR_045773.1 NR_045774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5887661 c.976C>T p.P326S Chr.2: 175097446-175097446 on Build GRCh38
4086885 c.944T>A p.L315* Chr.2: 175097478-175097478 on Build GRCh38
5649901 c.977C>T p.P326L Chr.2: 175097445-175097445 on Build GRCh38
1401403 c.935C>T p.P312L Chr.2: 175097487-175097487 on Build GRCh38
6606509 c.936G>A p.P312P Chr.2: 175097486-175097486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748186508 Chr.2: 175097465-175097465 on Build GRCh38
rs909827831 Chr.2: 175097308-175097308 on Build GRCh38
rs770319288 Chr.2: 175097444-175097444 on Build GRCh38
rs950481293 Chr.2: 175097317-175097317 on Build GRCh38
rs759250737 Chr.2: 175097458-175097458 on Build GRCh38
rs149284771 Chr.2: 175097377-175097377 on Build GRCh38
rs769608710 Chr.2: 175097468-175097468 on Build GRCh38
rs1006688344 Chr.2: 175097464-175097464 on Build GRCh38
rs757672996 Chr.2: 175097414-175097414 on Build GRCh38
rs990728981 Chr.2: 175097386-175097386 on Build GRCh38
rs1018823632 Chr.2: 175097447-175097447 on Build GRCh38
rs754443197 Chr.2: 175097413-175097413 on Build GRCh38
rs763407744 Chr.2: 175097367-175097370 on Build GRCh38
rs752901711 Chr.2: 175097395-175097395 on Build GRCh38
rs983686770 Chr.2: 175097361-175097361 on Build GRCh38
rs756295312 Chr.2: 175097410-175097410 on Build GRCh38
rs749300695 Chr.2: 175097388-175097388 on Build GRCh38
rs768206343 Chr.2: 175097446-175097446 on Build GRCh38
rs189365233 Chr.2: 175097272-175097272 on Build GRCh38
rs79006228 Chr.2: 175097347-175097347 on Build GRCh38
rs56300825 Chr.2: 175097445-175097445 on Build GRCh38
rs1045276427 Chr.2: 175097354-175097354 on Build GRCh38
rs578103084 Chr.2: 175097284-175097284 on Build GRCh38
rs758727345 Chr.2: 175097432-175097432 on Build GRCh38
rs139638515 Chr.2: 175097310-175097310 on Build GRCh38
rs934771554 Chr.2: 175097470-175097470 on Build GRCh38
rs376088944 Chr.2: 175097415-175097415 on Build GRCh38
rs1153682 Chr.2: 175097258-175097258 on Build GRCh38
rs764223377 Chr.2: 175097411-175097411 on Build GRCh38
rs779683235 Chr.2: 175097440-175097440 on Build GRCh38
rs370482532 Chr.2: 175097423-175097423 on Build GRCh38
rs563786319 Chr.2: 175097376-175097376 on Build GRCh38

View Full Product Details