Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97724378-97724603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
UniGene
Hs.234569
Gene Chromosome Location
Chr.2: 97712030-97744327 on Build GRCh38
Transcripts
NM_001079.3 XM_017004867.1 XM_017004868.1 XM_017004869.1 XM_017004870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5508801 c.360C>T p.D120D Chr.2: 97724396-97724396 on Build GRCh38
4618272 c.370C>T p.R124C Chr.2: 97724406-97724406 on Build GRCh38
3426776 c.382C>T p.R128C Chr.2: 97724418-97724418 on Build GRCh38
1410134 c.355C>T p.R119* Chr.2: 97724391-97724391 on Build GRCh38
6509586 c.346G>A p.D116N Chr.2: 97724382-97724382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767294957 Chr.2: 97724482-97724482 on Build GRCh38
rs371153389 Chr.2: 97724471-97724471 on Build GRCh38
rs919324397 Chr.2: 97724547-97724547 on Build GRCh38
rs560210342 Chr.2: 97724527-97724527 on Build GRCh38
rs200107228 Chr.2: 97724441-97724441 on Build GRCh38
rs925144320 Chr.2: 97724463-97724463 on Build GRCh38
rs868316208 Chr.2: 97724572-97724572 on Build GRCh38
rs201551373 Chr.2: 97724405-97724405 on Build GRCh38
rs768492740 Chr.2: 97724396-97724396 on Build GRCh38
rs545129970 Chr.2: 97724451-97724451 on Build GRCh38
rs199996419 Chr.2: 97724408-97724408 on Build GRCh38
rs768332464 Chr.2: 97724486-97724486 on Build GRCh38
rs867787314 Chr.2: 97724506-97724506 on Build GRCh38
rs1049218289 Chr.2: 97724568-97724568 on Build GRCh38
rs55824472 Chr.2: 97724465-97724465 on Build GRCh38
rs774255492 Chr.2: 97724539-97724539 on Build GRCh38
rs1040401534 Chr.2: 97724503-97724503 on Build GRCh38
rs777031626 Chr.2: 97724426-97724426 on Build GRCh38
rs996968760 Chr.2: 97724540-97724540 on Build GRCh38
rs200514384 Chr.2: 97724413-97724413 on Build GRCh38
rs750345154 Chr.2: 97724487-97724487 on Build GRCh38
rs201910784 Chr.2: 97724468-97724468 on Build GRCh38
rs549714930 Chr.2: 97724566-97724566 on Build GRCh38
rs770900178 Chr.2: 97724496-97724496 on Build GRCh38
rs747379748 Chr.2: 97724407-97724407 on Build GRCh38
rs774131317 Chr.2: 97724406-97724406 on Build GRCh38
rs757033990 Chr.2: 97724481-97724481 on Build GRCh38
rs929429105 Chr.2: 97724562-97724562 on Build GRCh38
rs765313500 Chr.2: 97724444-97724444 on Build GRCh38

View Full Product Details