Gene Transcripts

Gene Symbol
Fabp7
Entrez Gene ID
12140
Gene Name
fatty acid binding protein 7, brain
Gene Aliases
B-FABP, BFABP, Blbp, MRG
Chromosome Location
Chr. 10: 57784923 - 57788450 on Build GRCm38
UniGene ID
Mm.3644
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021272.3 NP_067247.1 3, 4 426
GenBank mRNA AJ870970.1 - 3, 4 426
AK002955.1 - 3, 4 427
AK021271.1 - 3, 4 431
BC055280.1 - 3, 4 409 5710017
BC057090.1 - 3, 4 412 5700428
CT010401.1 - 3, 4 340
S69799.1 - 3, 4 409

View Full Product Details