Gene Transcripts
- Gene Symbol
- Cap1
- Entrez Gene ID
- 12331
- Gene Name
- CAP, adenylate cyclase-associated protein 1 (yeast)
- Gene Aliases
- Chromosome Location
- Chr. 4: 122859048 - 122886071 on Build GRCm38
- UniGene ID
- Mm.8687
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301067.1 |
NP_001287996.1
|
8, 9 |
1052 |
|
NM_007598.4 |
NP_031624.2
|
8, 9 |
1036 |
|
GenBank mRNA |
AK053351.1 |
- |
8, 9 |
860 |
|
AK128889.1 |
- |
8, 9 |
858 |
|
AK137222.1 |
- |
8, 9 |
841 |
|
AK144094.1 |
- |
8, 9 |
844 |
|
AK154002.1 |
- |
8, 9 |
1056 |
|
AK156556.1 |
- |
8, 9 |
870 |
|
AK158150.1 |
- |
7, 8 |
1700 |
|
AK165565.1 |
- |
8, 9 |
839 |
|
AK169652.1 |
- |
8, 9 |
831 |
|
AK170901.1 |
- |
8, 9 |
847 |
|
AK171388.1 |
- |
8, 9 |
854 |
|
BC005446.1 |
- |
7, 8 |
835 |
3600514 |
BC005472.1 |
- |
7, 8 |
828 |
3590890 |
L12367.1 |
- |
8, 9 |
862 |
|
View Full Product Details