Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009892.2 | NP_034022.2 | 384 | ||
GenBank mRNA | AK137503.1 | - | 361 | ||
AK154420.1 | - | 359 | |||
BC061154.1 | - | 359 | 30298306 | ||
D87757.2 | - | 5 | 341 | ||
M94584.2 | - | 353 |