Gene Transcripts
- Gene Symbol
- Strc
- Entrez Gene ID
- 140476
- Gene Name
- stereocilin
- Gene Aliases
- DFNB16
- Chromosome Location
- Chr. 2: 121363248 - 121381370 on Build GRCm38
- UniGene ID
- Mm.467475
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080459.2 |
NP_536707.2
|
20 |
4203 |
|
GenBank mRNA |
AF375593.1 |
- |
20 |
4203 |
|
View Full Product Details