Gene Transcripts

Gene Symbol
Kcnj4
Entrez Gene ID
16520
Gene Name
potassium inwardly-rectifying channel, subfamily J, member 4
Gene Aliases
IRK3, Kcnf2, MB-IRK3
Chromosome Location
Chr. 15: 79483714 - 79511883 on Build GRCm38
UniGene ID
Mm.140760
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008427.4 NP_032453.3 2 430
GenBank mRNA AB024304.1 - 1 315
BC120593.1 - 2 407 40129516
BC137594.1 - 2 407 8860613
S71382.1 - 2 431
U11075.1 - 2 400

View Full Product Details