Gene Transcripts

Gene Symbol
Ppargc1b
Entrez Gene ID
170826
Gene Name
peroxisome proliferative activated receptor, gamma, coactivator 1 beta
Gene Aliases
4631412G21Rik, Perc
Chromosome Location
Chr. 18: 61291445 - 61401462 on Build GRCm38
UniGene ID
Mm.415302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133249.2 NP_573512.1 5 714
GenBank mRNA AF453324.1 - 5 714
AK028464.1 - 6 764
AK086111.1 - 5 713
BC150699.1 - 5 694 9087610

View Full Product Details