Gene Transcripts

Gene Symbol
Vmn1r234
Entrez Gene ID
171232
Gene Name
vomeronasal 1 receptor 234
Gene Aliases
V1rf1
Chromosome Location
Chr. 17: 21228826 - 21229815 on Build GRCm38
UniGene ID
Mm.377189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134198.1 NP_598959.1 1 568
GenBank mRNA BC127026.1 - 1 568

View Full Product Details