Gene Transcripts

Gene Symbol
Plxna3
Entrez Gene ID
18846
Gene Name
plexin A3
Gene Aliases
PlexA3, Plxa3, Plxn3, Plxn4, SEX, mKIAA4078
Chromosome Location
Chr. X: 74329066 - 74344689 on Build GRCm38
UniGene ID
Mm.1685
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008883.2 NP_032909.2 2 679
GenBank mRNA BC093482.1 - 2 662
D86950.1 - 2 645

View Full Product Details