Gene Transcripts
- Gene Symbol
- Slc4a1
- Entrez Gene ID
- 20533
- Gene Name
- solute carrier family 4 (anion exchanger), member 1
- Gene Aliases
- Ae1, CD233, Empb3, l11Jus51
- Chromosome Location
- Chr. 11: 102348820 - 102365281 on Build GRCm38
- UniGene ID
- Mm.7248
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011403.2 |
NP_035533.1
|
16, 17 |
2412 |
|
GenBank mRNA |
AK137542.1 |
- |
16, 17 |
2308 |
|
AK157833.1 |
- |
16, 17 |
2270 |
|
AK158184.1 |
- |
16, 17 |
2378 |
|
AK158243.1 |
- |
16, 17 |
2282 |
|
AK158453.1 |
- |
16, 17 |
2257 |
|
AK162616.1 |
- |
16, 17 |
2257 |
|
AK165195.1 |
- |
16, 17 |
2269 |
|
AK165267.1 |
- |
16, 17 |
2276 |
|
AK171662.1 |
- |
|
2308 |
|
AK171808.1 |
- |
16, 17 |
2246 |
|
AK171845.1 |
- |
|
2308 |
|
AK172103.1 |
- |
16, 17 |
2269 |
|
AK172316.1 |
- |
16, 17 |
2257 |
|
AK172395.1 |
- |
16, 17 |
2308 |
|
AK172456.1 |
- |
16, 17 |
2292 |
|
AK172569.1 |
- |
16, 17 |
2257 |
|
AY296129.1 |
- |
16, 17 |
2245 |
|
BC052419.1 |
- |
16, 17 |
2252 |
5702599 |
BC053429.1 |
- |
16, 17 |
2243 |
6506270 |
M29379.1 |
- |
16, 17 |
2239 |
|
X02677.1 |
- |
16, 17 |
2245 |
|
X03917.1 |
- |
14, 15 |
2089 |
|
View Full Product Details