Gene Transcripts

Gene Symbol
4930486L24Rik
Entrez Gene ID
214639
Gene Name
RIKEN cDNA 4930486L24 gene
Gene Aliases
AW545031
Chromosome Location
Chr. 13: 60842612 - 60864416 on Build GRCm38
UniGene ID
Mm.19839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178098.2 NP_835199.1 2 192
GenBank mRNA AK015635.1 - 2 162
AK145933.1 - 2 193
AY146988.1 - 2 192
BC061218.1 - 2 179 30281034

View Full Product Details