Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009464.3 | NP_033490.1 | 7 | 1080 | |
GenBank mRNA | AB008216.1 | - | 6 | 883 | |
AB010742.1 | - | 7 | 1080 | ||
AF030164.1 | - | 6 | 921 | ||
AF032902.1 | - | 7 | 1047 | ||
AF053352.1 | - | 7 | 1052 | ||
BC055901.1 | - | 6 | 1270 | ||
BC098234.1 | - | 5 | 1776 | ||
BC139430.1 | - | 6 | 948 | 8862452 | |
BC139431.1 | - | 6 | 948 | 8862453 |