Gene Transcripts
- Gene Symbol
- Camk1d
- Entrez Gene ID
- 227541
- Gene Name
- calcium/calmodulin-dependent protein kinase ID
- Gene Aliases
- A630059D12Rik, CKLiK, CaMKIdelta, E030025C11Rik, mCKLiK
- Chromosome Location
- Chr. 2: 5293457 - 5715364 on Build GRCm38
- UniGene ID
- Mm.191949
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290375.1 |
NP_001277304.1
|
2, 3 |
747 |
|
NM_001290376.1 |
NP_001277305.1
|
2, 3 |
263 |
|
NM_177343.4 |
NP_796317.2
|
2, 3 |
747 |
|
GenBank mRNA |
AK052401.1 |
- |
2, 3 |
168 |
|
AK053173.1 |
- |
2, 3 |
501 |
|
AK147616.1 |
- |
2, 3 |
748 |
|
AK147657.1 |
- |
2, 3 |
458 |
|
AK154434.1 |
- |
2, 3 |
336 |
|
AK162754.1 |
- |
2, 3 |
462 |
|
AK170777.1 |
- |
2, 3 |
512 |
|
AY273822.1 |
- |
2, 3 |
263 |
|
BC141413.1 |
- |
2, 3 |
247 |
9056026 |
BC141414.1 |
- |
2, 3 |
247 |
9056027 |
View Full Product Details