Gene Transcripts
- Gene Symbol
- Ppig
- Entrez Gene ID
- 228005
- Gene Name
- peptidyl-prolyl isomerase G (cyclophilin G)
- Gene Aliases
- AU019516, AU022200, B230312B02Rik, CYP, SRCyp
- Chromosome Location
- Chr. 2: 69722617 - 69754059 on Build GRCm38
- UniGene ID
- Mm.11815
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081086.1 |
NP_001074555.1
|
3, 4 |
429 |
|
GenBank mRNA |
AK088007.1 |
- |
3, 4 |
240 |
|
AK134121.1 |
- |
3, 4 |
431 |
|
AK135178.1 |
- |
4, 5 |
561 |
|
BC016126.1 |
- |
3, 4 |
291 |
|
BC027420.1 |
- |
3, 4 |
423 |
|
BC038818.1 |
- |
3, 4 |
247 |
|
BC048905.1 |
- |
3, 4 |
290 |
|
BC094926.1 |
- |
3, 4 |
283 |
|
BC150694.1 |
- |
4, 5 |
450 |
9087605 |
View Full Product Details