Gene Transcripts

Gene Symbol
Ppig
Entrez Gene ID
228005
Gene Name
peptidyl-prolyl isomerase G (cyclophilin G)
Gene Aliases
AU019516, AU022200, B230312B02Rik, CYP, SRCyp
Chromosome Location
Chr. 2: 69722617 - 69754059 on Build GRCm38
UniGene ID
Mm.11815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081086.1 NP_001074555.1 14 2303
GenBank mRNA AK164670.1 - 1 298
BC058789.1 - 1 333
BC150694.1 - 15 2324 9087605

View Full Product Details