Gene Transcripts

Gene Symbol
Fam124b
Entrez Gene ID
241128
Gene Name
family with sequence similarity 124, member B
Gene Aliases
A830043J08Rik
Chromosome Location
Chr. 1: 80186969 - 80218504 on Build GRCm38
UniGene ID
Mm.145511
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173425.3 NP_775601.1 2 1395
GenBank mRNA AK043867.1 - 2 1396
BC150872.1 - 2 1280 9087783

View Full Product Details