Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290784.1 | NP_001277713.1 | 4 | 528 | |
NM_178886.3 | NP_849217.2 | 4 | 531 | ||
GenBank mRNA | AK029683.1 | - | 4 | 429 | |
AK032208.1 | - | 4 | 417 | ||
AK184960.1 | - | 2 | 124 | ||
AK198364.1 | - | 2 | 129 | ||
AK202757.1 | - | 2 | 140 | ||
BC138177.1 | - | 4 | 531 | 8861196 | |
BC138178.1 | - | 5 | 529 | 8861197 | |
BC145174.1 | - | 4 | 527 | 9053725 | |
CN527209.1 | - | 4 | 448 |