Gene Transcripts

Gene Symbol
Wdr17
Entrez Gene ID
244484
Gene Name
WD repeat domain 17
Gene Aliases
3010002I12Rik, B230207L18Rik
Chromosome Location
Chr. 8: 54629611 - 54724517 on Build GRCm38
UniGene ID
Mm.95281
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172152.1 NP_001165623.1 7 1132
NM_028220.3 NP_082496.3 8 1155
GenBank mRNA AK044241.1 - 7 1133
BC028268.1 - 8 1166 5366098

View Full Product Details