Gene Transcripts

Gene Symbol
Olfr1009
Entrez Gene ID
258565
Gene Name
olfactory receptor 1009
Gene Aliases
MOR175-3
Chromosome Location
Chr. 2: 85721407 - 85722351 on Build GRCm38
UniGene ID
Mm.377570
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146572.2 NP_666783.1 1 115
GenBank mRNA BC111578.1 - 1 114
BC148238.1 - 1 115 40042413

View Full Product Details