Gene Transcripts

Gene Symbol
Zfp346
Entrez Gene ID
26919
Gene Name
zinc finger protein 346
Gene Aliases
Jaz, Znf346
Chromosome Location
Chr. 13: 55105271 - 55135071 on Build GRCm38
UniGene ID
Mm.22795
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012017.2 NP_036147.1 5 665
GenBank mRNA AF083339.1 - 5 663
AK185041.1 - 3 331
BC055075.1 - 5 659 30056643
CV562260.1 - 4 461

View Full Product Details