Gene Transcripts

Gene Symbol
Rabep1
Entrez Gene ID
54189
Gene Name
rabaptin, RAB GTPase binding effector protein 1
Gene Aliases
Rab5ep, rabaptin-5, rabaptin-5alpha
Chromosome Location
Chr. 11: 70844723 - 70943105 on Build GRCm38
UniGene ID
Mm.7087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291141.1 NP_001278070.1 13, 14 2303
NM_001291142.1 NP_001278071.1 13, 14 2312
NM_001291143.1 NP_001278072.1 15, 16 2503
NM_019400.3 NP_062273.2 14, 15 2432
GenBank mRNA AB001750.1 - 15, 16 2339
AF248489.1 - 13, 14 2098
AF248490.1 - 13, 14 2089
AK134587.1 - 14, 15 2433
BC003921.1 - 5, 6 1053
BC080274.1 - 14, 15 2404
BC129854.1 - 14, 15 2251 40109689
D86066.1 - 14, 15 2268

View Full Product Details