Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020560.2 | NP_065585.1 | 1 | 139 | |
GenBank mRNA | AK005110.1 | - | 1 | 98 | |
AK011825.1 | - | 1 | 95 | ||
AK078073.1 | - | 1 | 82 | ||
AK161913.1 | - | 1 | 82 | ||
BC006764.1 | - | 1 | 34 | ||
BC018475.1 | - | 1 | 38 | 3492633 | |
BC027430.1 | - | 1 | 43 | 4187438 | |
BY237441.1 | - | 1 | 141 | ||
Z46966.1 | - | 1 | 58 |