Gene Transcripts

Gene Symbol
Nacc1
Entrez Gene ID
66830
Gene Name
nucleus accumbens associated 1, BEN and BTB (POZ) domain containing
Gene Aliases
2010001H03Rik, 4930511N13Rik, Btbd14b, Nac1
Chromosome Location
Chr. 8: 84670476 - 84687985 on Build GRCm38
UniGene ID
Mm.289698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025788.3 NP_080064.3 6 1727
GenBank mRNA AK012935.1 - 6 1710
AK197471.1 - 1 69
AK217624.1 - 1 69
BC048157.1 - 6 1700 5697941
BC052706.1 - 6 1710 5702150
BC054800.1 - 6 1727 6850337

View Full Product Details