Gene Transcripts

Gene Symbol
Ilkap
Entrez Gene ID
67444
Gene Name
integrin-linked kinase-associated serine/threonine phosphatase 2C
Gene Aliases
0710007A14Rik, 1600009O09Rik, PP2C-DELTA
Chromosome Location
Chr. 1: 91373846 - 91398841 on Build GRCm38
UniGene ID
Mm.337240
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023343.2 NP_075832.1 4, 5 373
GenBank mRNA BC011427.1 - 4, 5 343
BC026953.1 - 4, 5 373 5133980

View Full Product Details