Gene Transcripts

Gene Symbol
Dym
Entrez Gene ID
69190
Gene Name
dymeclin
Gene Aliases
1810041M12Rik, 4933427L07Rik, C030019K18Rik
Chromosome Location
Chr. 18: 75018699 - 75286966 on Build GRCm38
UniGene ID
Mm.173826
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027727.2 NP_082003.1 4 500
GenBank mRNA AK016957.1 - 4 429
AK049909.1 - 4 465
AK080149.1 - 4 477
AK081242.1 - 4 492
AK149628.1 - 4 474
AK149879.1 - 4 475
AK149891.1 - 4 474
AK154794.1 - 4 489
AK199364.1 - 2 112
BC038276.1 - 4 502 4458792
BC092283.1 - 4 486

View Full Product Details