Gene Transcripts

Gene Symbol
Med26
Entrez Gene ID
70625
Gene Name
mediator complex subunit 26
Gene Aliases
5730493L18Rik, AI414941, AW495270, Crsp7
Chromosome Location
Chr. 8: 72493453 - 72548310 on Build GRCm38
UniGene ID
Mm.235885
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027485.4 NP_081761.2 3 1440
GenBank mRNA AK017726.1 - 1 464
AK083035.1 - 3 1401
BC054737.1 - 3 1168 6401737

View Full Product Details