Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028450.3 | NP_082726.2 | 6 | 589 | |
GenBank mRNA | AK014093.1 | - | 5 | 338 | |
AK017798.1 | - | 6 | 567 | ||
AK028490.1 | - | 5 | 505 | ||
AK036670.1 | - | 6 | 589 | ||
BC032154.1 | - | 6 | 760 | 5068339 | |
BY713087.1 | - | 5 | 336 |