Gene Transcripts

Gene Symbol
Phf10
Entrez Gene ID
72057
Gene Name
PHD finger protein 10
Gene Aliases
1810055P05Rik, AV024533
Chromosome Location
Chr. 17: 14944994 - 14961638 on Build GRCm38
UniGene ID
Mm.440085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024250.4 NP_077212.3 6 724
GenBank mRNA AB221576.1 - 4 391
AK007873.1 - 6 704
BC002206.1 - 4 458 3489870
CA322732.1 - 3 282

View Full Product Details