Gene Transcripts

Gene Symbol
Spink5
Entrez Gene ID
72432
Gene Name
serine peptidase inhibitor, Kazal type 5
Gene Aliases
2310065D10Rik, AV238912, LEKT1, LETK1, VAKT1
Chromosome Location
Chr. 18: 43959545 - 44022487 on Build GRCm38
UniGene ID
Mm.35369
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081180.1 NP_001074649.1 31 3058
GenBank mRNA AJ426449.2 - 31 3056
AK028727.1 - 12 1208
BC118011.1 - 31 3030 40097427

View Full Product Details