Gene Transcripts

Gene Symbol
Ddias
Entrez Gene ID
74041
Gene Name
DNA damage-induced apoptosis suppressor
Gene Aliases
4632434I11Rik, noxin
Chromosome Location
Chr. 7: 92857525 - 92874286 on Build GRCm38
UniGene ID
Mm.26468
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080995.1 NP_001074464.1 7 1402
GenBank mRNA AK154915.1 - 6 1306
BC066148.1 - 6 1296 30086532
DQ400346.1 - 6 1336

View Full Product Details