Gene Transcripts

Gene Symbol
Morc2a
Entrez Gene ID
74522
Gene Name
microrchidia 2A
Gene Aliases
8430403M08Rik, Zcwcc1
Chromosome Location
Chr. 11: 3649494 - 3690477 on Build GRCm38
UniGene ID
Mm.139127
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159288.1 NP_001152760.1 11 1614
NM_198162.1 NP_937805.1 10 941
GenBank mRNA AK173042.1 - 11 1614
BC059243.1 - 10 941 6417551

View Full Product Details