Gene Transcripts

Gene Symbol
4930502E18Rik
Entrez Gene ID
75013
Gene Name
RIKEN cDNA 4930502E18 gene
Gene Aliases
Ct55
Chromosome Location
Chr. X: 53724831 - 53738565 on Build GRCm38
UniGene ID
Mm.336332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029142.1 NP_083418.1 4, 5 642
GenBank mRNA AK015678.1 - 4, 5 648
BC115663.1 - 4, 5 642 40103303
BC115664.1 - 4, 5 642 40103304

View Full Product Details