Gene Transcripts

Gene Symbol
0610040J01Rik
Entrez Gene ID
76261
Gene Name
RIKEN cDNA 0610040J01 gene
Gene Aliases
AI662686
Chromosome Location
Chr. 5: 63811969 - 63899626 on Build GRCm38
UniGene ID
Mm.38143
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029554.4 NP_083830.3 3 191
GenBank mRNA AK002869.1 - 3 192
AK050792.1 - 3 323
AK159779.1 - 3 154
BC004797.1 - 3 108 3589105

View Full Product Details