Gene Transcripts

Gene Symbol
Sowahb
Entrez Gene ID
78088
Gene Name
sosondowah ankyrin repeat domain family member B
Gene Aliases
5730467H21Rik, Ankrd56
Chromosome Location
Chr. 5: 93041123 - 93045022 on Build GRCm38
UniGene ID
Mm.245085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175270.4 NP_780479.1 1 2186
GenBank mRNA AK033423.1 - 1 2116
AK033641.1 - 1 1649
AK137429.1 - 1 2187
AK144982.1 - 1 230

View Full Product Details