Gene Transcripts

Gene Symbol
Mrps35
Entrez Gene ID
232536
Gene Name
mitochondrial ribosomal protein S35
Gene Aliases
MDSO23, MRP-S28, MRPS28
Chromosome Location
Chr. 6: 147042741 - 147070903 on Build GRCm38
UniGene ID
Mm.46656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145573.2 NP_663548.2 5 468
GenBank mRNA AK077677.1 - 5 469
AK170480.1 - 5 468
BC019964.1 - 5 446 4158657

View Full Product Details