Gene Transcripts

Gene Symbol
Olfr1419
Entrez Gene ID
257938
Gene Name
olfactory receptor 1419
Gene Aliases
MOR266-10
Chromosome Location
Chr. 19: 11870183 - 11871214 on Build GRCm38
UniGene ID
Mm.377309
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011775.1 NP_001011775.1 1 894
GenBank mRNA BC147200.1 - 1 985
BC147201.1 - 1 985 9056264

View Full Product Details