Gene Transcripts

Gene Symbol
Olfr873
Entrez Gene ID
258554
Gene Name
olfactory receptor 873
Gene Aliases
MOR145-2
Chromosome Location
Chr. 9: 20300211 - 20301170 on Build GRCm38
UniGene ID
Mm.334991
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146561.1 NP_666772.1 1 109
GenBank mRNA BC139140.1 - 1 192

View Full Product Details