Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023065.3 | NP_075552.2 | 2 | 260 | |
GenBank mRNA | AF290973.1 | - | 2 | 261 | |
AF309649.1 | - | 2 | 228 | ||
AK077725.1 | - | 2 | 261 | ||
AK156914.1 | - | 2 | 261 | ||
BC054852.1 | - | 2 | 246 | 6528891 | |
BQ746455.1 | - | 2 | 215 | ||
BY774497.1 | - | 2 | 261 |