Gene Transcripts

Gene Symbol
Nipsnap3b
Entrez Gene ID
66536
Gene Name
nipsnap homolog 3B (C. elegans)
Gene Aliases
2700063N13Rik, Nipsnap3a
Chromosome Location
Chr. 4: 53011775 - 53022060 on Build GRCm38
UniGene ID
Mm.38244
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025623.2 NP_079899.1 2 159
GenBank mRNA AK004458.1 - 2 158
AK008786.1 - 2 155
AK012480.1 - 2 159
BC019505.1 - 2 144 4164310

View Full Product Details